MilliporeSigma
  • Selective agonists of NK-2 binding sites highly active on rat portal vein (NK-3 bioassay).
Selective agonists of NK-2 binding sites highly active on rat portal vein (NK-3 bioassay).
Neuropeptides (1991-06-01)
G Chassaing, S Lavielle, D Loeuillet, P Robilliard, A Carruette, C Garret, J C Beaujouan, M Saffroy, F Petitet, Y Torrens
ABSTRACT
All the synthetized NKA and NKA (4-10) agonists have been found active in the rat portal vein bioassay. Even [Lys5, MeLeu9, Nle10] NKA(4-10), a highly potent competitor of NK-2 binding sites with very low binding potencies for NK-1 and NK-3 sites (IC50 greater than microM) is still active in contracting the rat portal vein. These results suggest that this tissue contains not only a fairly large population of NK-3 receptors but also a minor population of NK-2 receptors. Comparison of the activities of NKA C-terminal analogues on the guinea-pig ileum suggests that 1) only a small population of NK-2 receptors are present in this tissue and 2) beside NK-1, NK-2 and NK-3 receptors, another type of receptor sensitive to C-terminal sequences might be present in the guinea-pig tissue.