MilliporeSigma
  • Potentiometric detection of ATP based on the transmembrane proton gradient generated by ATPase reconstituted on a gold electrode.
Potentiometric detection of ATP based on the transmembrane proton gradient generated by ATPase reconstituted on a gold electrode.
Bioelectrochemistry (Amsterdam, Netherlands) (2020-03-04)
Gabriel García-Molina, Paolo Natale, Laura Valenzuela, Julia Alvarez-Malmagro, Cristina Gutiérrez-Sánchez, Ana Iglesias-Juez, Iván López-Montero, Marisela Vélez, Marcos Pita, Antonio L De Lacey
ABSTRACT
Adenosine triphosphate (ATP) is a key molecule as energy vector for living organisms, therefore its detection reveals the presence of microbial colonies. Environments where the existence of microbial pathogens suppose a health hazard can benefit from real time monitoring of such molecule. We report a potentiometric biosensor based on ATP-synthase from Escherichia coli reconstituted in a floating phospholipid bilayer over gold electrodes modified with a 4-aminothiophenol self-assembled monolayer. The use of a pH-dependent redox probe on the electrode surface allows a simple, specific and reliable on site determination of ATP concentration from 1 μM to 1 mM. The broad range ATP biosensor can offer an alternative way of measuring in a few minutes the presence of microbial contamination.
Sigma-Aldrich
4-Aminothiophenol, ≥97.0%