MilliporeSigma
  • An internal EELD domain facilitates mitochondrial targeting of Mcl-1 via a Tom70-dependent pathway.
An internal EELD domain facilitates mitochondrial targeting of Mcl-1 via a Tom70-dependent pathway.
Molecular biology of the cell (2006-07-11)
Chiang-Hung Chou, Ru-Shuo Lee, Hsin-Fang Yang-Yen
ABSTRACT
Mcl-1 functions at an apical step in many regulatory programs that control cell death. Although the mitochondrion is one major subcellular organelle where Mcl-1 functions, the molecular mechanism by which Mcl-1 is targeted to mitochondria remains unclear. Here, we demonstrate that Mcl-1 is loosely associated with the outer membrane of mitochondria. Furthermore, we demonstrate that Mcl-1 interacts with the mitochondrial import receptor Tom70, and such interaction requires an internal domain of Mcl-1 that contains an EELD motif. A Tom70 antibody that blocks Mcl-1-Tom70 interaction blocks mitochondrial import of Mcl-1 in vitro. Furthermore, Mcl-1 is significantly less targeted to mitochondria in Tom70 knockdown than in the control cells. Similar targeting preference is also observed for the DM mutant of Mcl-1 whose mutation at the EELD motif markedly attenuates its Tom70 binding activity. Together, our results indicate that the internal EELD domain facilitates mitochondrial targeting of Mcl-1 via a Tom70-dependent pathway.