MilliporeSigma
  • Neuron-Specific Tumor Necrosis Factor Receptor-Associated Factor 3 Is a Central Regulator of Neuronal Death in Acute Ischemic Stroke.
Neuron-Specific Tumor Necrosis Factor Receptor-Associated Factor 3 Is a Central Regulator of Neuronal Death in Acute Ischemic Stroke.
Hypertension (Dallas, Tex. : 1979) (2015-08-14)
Jun Gong, Zuo-Zhi Li, Sen Guo, Xiao-Jing Zhang, Peng Zhang, Guang-Nian Zhao, Lu Gao, Yan Zhang, Ankang Zheng, Xiao-Fei Zhang, Mei Xiang, Hongliang Li
ABSTRACT
Neuronal death after ischemic stroke involves multiple pathophysiological events, as well as a complex molecular mechanism. Inhibiting a single therapeutic target that is involved in several ischemic signaling cascades may be a promising strategy for stroke management. Here, we report the versatile biological roles of tumor necrosis factor receptor-associated factor 3 (TRAF3) in ischemic stroke. Using several genetically manipulated mouse strains, we also demonstrated that TRAF3 inhibition can be neuroprotective. TRAF3 expression, which is robustly induced in response to ischemia/reperfusion (I/R) injury, was detected in neurons. Overexpression of TRAF3 in neurons led to aggravated neuronal loss and enlarged infarcts; these effects were reversed in TRAF3-knockout mice. Neuronal TRAF3 also contributed to c-Jun kinase-, nuclear factor κB- and Rac-1-induced neuronal death, inflammation, and oxidative stress. Mechanistically, we showed that TRAF3 interacts with transforming growth factor-β-activated kinase 1 (TAK1) and potentiates phosphorylation and activation of TAK1. Phosphorylated TAK1 sequentially initiated activation of nuclear factor κB, Rac-1/NADPH oxidase, and c-Jun kinase/c-Jun signaling cascades. Using a combination of adenoviruses encoding dominant-negative TAK1 and the TAK1 inhibitor 5Z-7-oxozeaenol, we demonstrated that the TRAF3-mediated activation of ischemic cascades was TAK1-dependent. More importantly, the adverse phenotypes observed in TRAF3-overexpressing mice were completely reversed when the TRAF3-TAK1 interaction was prevented. Therefore, we have shown that TRAF3 is a central regulator of ischemic pathways, including nuclear factor κB, Rac-1, and c-Jun kinase signaling, via its interaction with and activation of TAK1. Furthermore, certain components of the TRAF3-TAK1 signaling pathway are potentially promising therapeutic targets in ischemic stroke.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse