MilliporeSigma
  • MiR-708-5p is inversely associated with EWS/FLI1 Ewing sarcoma but does not represent a prognostic predictor.
MiR-708-5p is inversely associated with EWS/FLI1 Ewing sarcoma but does not represent a prognostic predictor.
Cancer genetics (2018-11-25)
Gabriela Molinari Roberto, Gabriela Maciel Vieira, Lara Elis Alberici Delsin, Marcela de Oliveira Silva, Rodrigo Guedes Hakime, Edgard Eduard Engel, Carlos Alberto Scrideli, Luiz Gonzaga Tone, María Sol Brassesco
ABSTRACT
Overall survival of Ewing sarcoma (EWS) remains poor and less than 30% of patients with metastatic or recurrent disease survive despite current treatments. Thus, there is a constant search for new biomarkers for diagnosis, prognosis and prediction of therapy. Numerous studies have reported the abnormal expression of miR-708-5p in tumors of different origins. However, its role in EWS remains unclear. qRT-PCR was performed in nineteen consecutive EWS samples and twelve non-tumor bone samples from age-matched controls. Functional assays were performed in SK-ES-1 cells transfected with miR-708 lentiviral-based vectors and results analyzed in terms of clonogenicity, migration, invasion and western blot. We show that miR-708-5p is downregulated in EWS tissues though no associations with any prognostic features such as HUVOS grade, event or survival were found in our cohort. Nonetheless, expression levels of this micro-RNA were inversely associated with the presence of the EWS/FLI1 translocation. When miR-708-5p was transfected into the SK-ES-1 cell line, it did not affect migration or clonogenicity, but promoted a significant increase on the invasive potential of cells endorsed with high expression of MMP2. Taken together, our results suggest that despite downregulated in EWS samples, this miRNA might represent a secondary genetic alteration derived from the pleiotropic cellular effects of the abnormal EWS/FLI1 transcription factor that does not affect tumor growth but instead, is related with the promotion of tumor invasion, not being suitable for future therapeutic intervention.