MilliporeSigma
  • Environmental and cellular factors affecting the localization of T6SS proteins in Burkholderia thailandensis.
Environmental and cellular factors affecting the localization of T6SS proteins in Burkholderia thailandensis.
International journal of medical microbiology : IJMM (2019-08-06)
Jan Lennings, Munira Makhlouf, Przemyslaw Olejnik, Christian Mayer, Heike Brötz-Oesterhelt, Sandra Schwarz
ABSTRACT
The type VI secretion system (T6SS) injects effector proteins into neighboring bacteria and host cells. Effector translocation is driven by contraction of a tubular sheath in the cytoplasm that expels an inner needle across the cell envelope. The AAA + ATPase ClpV disassembles and recycles the contracted sheath. While ClpV-1-GFP of the Burkholderia T6SS-1, which targets prokaryotic cells, assembles into randomly localized foci, ClpV-5-GFP of the virulence-associated T6SS-5 displays a polar distribution. The mechanisms underlying the localization of T6SSs to a particular site in the bacterial cell are currently unknown. We recently showed that ClpV-5-GFP retains its polar localization in the absence of all T6SS-5 components during infection of host cells. Herein, we set out to identify factors involved in the distribution of ClpV-5 and ClpV-1 in Burkholderia thailandensis. We show that focal assembly and polar localization of ClpV-5-GFP is not dependent on the intracellular host cell environment, known to contain the signal to induce T6SS-5 gene expression. In contrast to ClpV-5-GFP, localization of ClpV-1-GFP was dependent on the cognate T6SS. Foci formation of both ClpV5-GFP and ClpV-1-GFP was decreased by D cycloserine-mediated inhibition of peptidoglycan synthesis while treatment of B. thailandensis with A22 blocking the cytoskeletal protein MreB did not affect assembly of ClpV-5 and ClpV-1 into single discrete foci. Furthermore, we found that surface contact promotes but is not essential for localization of ClpV-5-GFP to the pole whereas expression of clpV-1-gfp appears to be induced by surface contact. In summary, the study provides novel insights into the localization of ClpV ATPases of T6SSs targeting prokaryotic and eukaryotic cells.
Sigma-Aldrich
A22 hydrochloride, ≥95% (HPLC)