MilliporeSigma
  • MicroRNA-582-3p negatively regulates cell proliferation and cell cycle progression in acute myeloid leukemia by targeting cyclin B2.
MicroRNA-582-3p negatively regulates cell proliferation and cell cycle progression in acute myeloid leukemia by targeting cyclin B2.
Cellular & molecular biology letters (2019-12-18)
Haixia Li, Xuefei Tian, Paoqiu Wang, Mao Huang, Ronghua Xu, Tian Nie
ABSTRACT
MicroRNAs (miRNAs) function as post-transcriptional gene expression regulators. Some miRNAs, including the recently discovered miR-582-3p, have been implicated in leukemogenesis. This study aimed to reveal the biological function of miR-582-3p in acute myeloid leukemia (AML), which is one of the most frequently diagnosed hematological malignancies. The expression of miR-582-3p was determined using quantitative real-time PCR in blood samples from leukemia patients and in cell lines. Cell proliferation and cell cycle distribution were analyzed using the CCK-8, colony formation and flow cytometry assays. The target gene of miR-582-3p was verified using a dual-luciferase reporter assay. The G2/M phase arrest-related molecule contents were measured using western blotting analysis. We found miR-582-3p was significantly downregulated in the blood samples from leukemia patients and in the cell lines. MiR-582-3p overexpression significantly impaired cell proliferation and induced G2/M cell cycle arrest in THP-1 cells. Furthermore, cyclin B2 (CCNB2) was confirmed as a target gene of miR-582-3p and found to be negatively regulated by miR-582-3p overexpression. More importantly, CCNB2 knockdown showed suppressive effects on cell proliferation and cell cycle progression similar to those caused by miR-582-3p overexpression. The inhibitory effects of miR-582-3p overexpression on cell proliferation and cell cycle progression were abrogated by CCNB2 transfection. These findings indicate new functions and mechanisms for miR-582-3p in AML development. Further study could clarify if miR-582-3p and CCNB2 are potential therapeutic targets for the treatment of AML.
Sigma-Aldrich
MISSION® esiRNA, targeting human CCNB2
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder