MilliporeSigma
  • Priming of transcriptional memory responses via the chromatin accessibility landscape in T cells.
Priming of transcriptional memory responses via the chromatin accessibility landscape in T cells.
Scientific reports (2017-03-21)
Wen Juan Tu, Kristine Hardy, Christopher R Sutton, Robert McCuaig, Jasmine Li, Jenny Dunn, Abel Tan, Vedran Brezar, Melanie Morris, Gareth Denyer, Sau Kuen Lee, Stephen J Turner, Nabila Seddiki, Corey Smith, Rajiv Khanna, Sudha Rao
ABSTRACT
Memory T cells exhibit transcriptional memory and "remember" their previous pathogenic encounter to increase transcription on re-infection. However, how this transcriptional priming response is regulated is unknown. Here we performed global FAIRE-seq profiling of chromatin accessibility in a human T cell transcriptional memory model. Primary activation induced persistent accessibility changes, and secondary activation induced secondary-specific opening of previously less accessible regions associated with enhanced expression of memory-responsive genes. Increased accessibility occurred largely in distal regulatory regions and was associated with increased histone acetylation and relative H3.3 deposition. The enhanced re-stimulation response was linked to the strength of initial PKC-induced signalling, and PKC-sensitive increases in accessibility upon initial stimulation showed higher accessibility on re-stimulation. While accessibility maintenance was associated with ETS-1, accessibility at re-stimulation-specific regions was linked to NFAT, especially in combination with ETS-1, EGR, GATA, NFκB, and NR4A. Furthermore, NFATC1 was directly regulated by ETS-1 at an enhancer region. In contrast to the factors that increased accessibility, signalling from bHLH and ZEB family members enhanced decreased accessibility upon re-stimulation. Interplay between distal regulatory elements, accessibility, and the combined action of sequence-specific transcription factors allows transcriptional memory-responsive genes to "remember" their initial environmental encounter.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, 0.8 mg/mL, Chemicon®