MilliporeSigma
  • Involvement of PERK-CHOP pathway in fumonisin B1- induced cytotoxicity in human gastric epithelial cells.
Involvement of PERK-CHOP pathway in fumonisin B1- induced cytotoxicity in human gastric epithelial cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-01-01)
Song Yu, Bingxuan Jia, Yunxia Yang, Na Liu, Aibo Wu
ABSTRACT
Fumonisin B1 (FB1) is a mycotoxin, produced by Fusarium verticillioides and Fusarium proliferatum, and a common fungal contaminant of maize worldwide. Its potential health hazard as a natural toxin is well documented in human and domestic animals. However, the molecular mechanism and the key factors responsible for FB1-induced cytotoxicity have not been elucidated. In this study, we first examined the cytotoxicity induced by FB1 in human gastric epithelial cell line (GES-1). We found that FB1 notably decreased cell viability and induced apoptotic cell death. Furthermore, the levels of ER stress markers were significantly increased after FB1 exposure and the ER stress inhibitor 4-phenylbutyric acid strongly suppressed FB1-induced cytotoxicity. Interestingly, the inhibition of PERK activity by GSK2606414 or shPERK3 blocked FB1-induced apoptotic cell death and cell proliferation suppression, which indicated that the cytotoxicity induced by FB1 was dependent on this signalling pathway. Moreover, myriocin could relieve FB1-induced ER stress and prevent cell death, which implied that the disruption of sphingolipid metabolism is an apical event for FB1-induced cytotoxicity. In the present study, we demonstrated that the ER stress-related PERK-CHOP signalling pathway is a novel mechanism for FB1-induced cytotoxicity and the gastrointestinal injury caused by FB1 should be concerned in the future.
Sigma-Aldrich
Myriocin, Mycelia sterilia, A potent immunosuppressant.
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder