- Time-dependent matrix metalloproteinases and tissue inhibitor of metalloproteinases expression change in fusarium solani keratitis.
Time-dependent matrix metalloproteinases and tissue inhibitor of metalloproteinases expression change in fusarium solani keratitis.
To investigate matrix metalloproteinases (MMPs) and tissue inhibitor of metalloproteinases (TIMPs) expression during the progress of fusarium solani (F.solani) keratitis in a rat model. A rat model of F.solani keratitis was produced using corneal scarification and a hand-made contact lens. MMPs and TIMPs expressiond were explored in this rat model of F.solani keratitis using real-time polymerase chain reaction (PCR) and DIF. GM6001 (400 µmol/mL) was used to treat infected corneas. The keratitis duration, amount and area of corneal neovascularization (CNV) were evaluated. MMP-3 expression was 66.3 times higher in infected corneas compared to normal corneas. MMP-8, -9, and -13 expressions were significantly upregulated in the mid-period of the infection, with infected-to-normal ratios of 4.03, 39.86, and 5.94, respectively. MMP-2 and -7 expressions increased in the late period, with the infected-to-normal ratios of 5.94 and 16.22, respectively. TIMP-1 expression was upregulated in the early period, and it was 43.17 times higher in infected compared to normal corneas, but TIMP-2, -3, and -4 expressions were mildly downregulated or unchanged. The results of DIF were consistent with the result of real-time PCR. GM6001, a MMPs inhibitor, decreased the duration of F.solani infection and the amount and area of CNV. MMPs and TIMPs contributed into the progress of F.solani keratitis.