MilliporeSigma
  • Fluorometric sensing of oxygen using manganese(II)-doped zinc sulfide nanocrystals.
Fluorometric sensing of oxygen using manganese(II)-doped zinc sulfide nanocrystals.
Mikrochimica acta (2019-12-20)
Fangyuan Lin, Zhiwei Lai, Linchun Zhang, Yipeng Huang, Feiming Li, Pingyun Chen, Yiru Wang, Xi Chen
ABSTRACT
Manganese(II)-doped zinc sulfide nanocrystals (Mn:ZnS NCs) with dual-emission fluorescence (peaks at 445 nm and 590 nm under 330 nm excitation), good water stability and low toxicity were synthesized by hot injection. The fluorescence intensity of both emission bands of the nanocrystals can change rapidly by the content of gaseous and dissolved oxygen. The process is fully reversible. Compared with the maximum intensity of Mn:ZnS sensing film in 100% nitrogen, the emission of the blue emission decreases by 72% in the presence of 100% oxygen, and the yellow emission by 32%. Response is linear in the presence of 3% to 12% of oxygen percentage in gas. For water-dissolved oxygen, the linear response occurs between 0.54 and 11.4 mg·L-1. Graphical abstractMn-doped ZnS NCs with dual-emission fluorescence were synthesized by hot-injection method. The reversible and rapid sensing characteristics of Mn-doped ZnS NCs to oxygen were studied, and the possible sensing mechanism was investigated.
Sigma-Aldrich
Sulfur, purum p.a., ≥99% (T)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)