MilliporeSigma
  • Molecular cloning and expression of the nucleoside deoxyribosyltransferase-II gene from Lactobacillus helveticus.
Molecular cloning and expression of the nucleoside deoxyribosyltransferase-II gene from Lactobacillus helveticus.
Bioscience, biotechnology, and biochemistry (2000-12-29)
K Okuyama, T Noguchi
ABSTRACT
Nucleoside deoxyribosyltransferase-II, which catalyzes transfer of glycosyl residues from a donor deoxynucleoside to an acceptor base, was purified from Lactobacillus helveticus and its gene was cloned. Analysis of the nucleotide sequence showed the presence of a 474-nucleotide open reading frame encoding a protein of 158 amino acids with a molecular weight of 18,317. The active enzyme can be produced in large quantities in E. coli cells using the cloned gene.