MilliporeSigma
  • Mutations in RNF135, a gene within the NF1 microdeletion region, cause phenotypic abnormalities including overgrowth.
Mutations in RNF135, a gene within the NF1 microdeletion region, cause phenotypic abnormalities including overgrowth.
Nature genetics (2007-07-17)
Jenny Douglas, Deirdre Cilliers, Kim Coleman, Katrina Tatton-Brown, Karen Barker, Brigitte Bernhard, John Burn, Susan Huson, Dragana Josifova, Didier Lacombe, Mohsin Malik, Sahar Mansour, Evan Reid, Valerie Cormier-Daire, Trevor Cole, Nazneen Rahman
ABSTRACT
17q11 microdeletions that encompass NF1 cause 5%-10% of cases of neurofibromatosis type 1, and individuals with microdeletions are typically taller than individuals with intragenic NF1 mutations, suggesting that deletion of a neighboring gene might promote human growth. We identified mutations in RNF135, which is within the NF1 microdeletion region, in six families characterized by overgrowth, learning disability, dysmorphic features and variable additional features. These data identify RNF135 as causative of a new overgrowth syndrome and demonstrate that RNF135 haploinsufficiency contributes to the phenotype of NF1 microdeletion cases.