MilliporeSigma
PAS kinase deficiency reduces aging effects in mice.
Aging (2020-01-25)
Pilar Dongil, Ana Pérez-García, Verónica Hurtado-Carneiro, Carmen Herrero-de-Dios, Elvira Álvarez, Carmen Sanz
ABSTRACT
Several signaling pathways may be affected during aging. All are regulated by nutrient levels leading to a decline in mitochondrial function and autophagy and to an increase in oxidative stress. PAS Domain Kinase (PASK) is a nutrient and bioenergetic sensor. We have previously found that PASK plays a role in the control of hepatic metabolic balance and mitochondrial homeostasis. To investigate PASK's role in hepatic oxidative stress during aging, we analyzed the mitochondrial function, glucose tolerance, insulin resistance, and lipid-related parameters in aged PASK-deficient mice. Hepatic Pask mRNA decreased in step with aging, being undetectable in aged wild-type (WT) mice. Aged PASK-deficient mice recorded lower levels of ROS/RNS compared to aged WT. The regulators of mitochondrial biogenesis, PGC1a, SIRT1 and NRF2, decreased in aged WT, while aged PASK-deficient mice recorded a higher expression of NRF2, GCLm and HO1 proteins and CS activity under fasted conditions. Additionally, aged PASK-deficient mice recorded an overexpression of the longevity gene FoxO3a, and maintained elevated PCNA protein, suggesting that hepatic cell repair mechanisms might be functional. PASK-deficient mice have better insulin sensitivity and no glucose intolerance, as confirmed by a normal HOMA-IR index. PASK may be a good target for reducing damage during aging.
Sigma-Aldrich
Citrate Synthase Activity Assay Kit, Sufficient for 100 Colorimetric tests