MilliporeSigma
  • Pectinase production by Neurospora crassa: purification and biochemical characterization of extracellular polygalacturonase activity
Pectinase production by Neurospora crassa: purification and biochemical characterization of extracellular polygalacturonase activity
Microbiology (1991)
Maria de Lourdes T. M. Polizeli, et al.