MilliporeSigma
  • A two-dimensional molecular network structure of trimesic acid prepared by adsorption-induced self-organization
A two-dimensional molecular network structure of trimesic acid prepared by adsorption-induced self-organization
Chemical Communications (Cambridge, England) (2002)
Ishikawa Y, et al.