MilliporeSigma
  • Analysis of surface properties of human cancer cells using derivatized beads.
Analysis of surface properties of human cancer cells using derivatized beads.
Acta histochemica (2002-10-23)
Maria R Khurrum, Gayani R Weerasinghe, Evelyn S Soriano, Rashad Riman, Oliver Badali, Stephanie Gipson, Jessica Medina, Juan Alfaro, Vanessa M Navarro, Caroline B Harieg, Lylla Ngo, Tharinee Sakhakorn, Lital Kirszenbaum, David Khatibi, Karolin Abedi, Marcela Barajas, Gregory C Zem, Adit Kirszenbaum, Arash Razi, Steven B Oppenheimer
ABSTRACT
Standard histochemical analysis of cells and tissues generally involves procedures that utilize a relatively small number of probes such as dyes, and generally requires hours or days to process. Our laboratory has developed a novel method for histochemical surveys of cell surface properties that utilizes a large number of probes (derivatized agarose beads) and takes seconds or minutes to accomplish. In this study, 4 human cell lines (CCL-255 (LS123) human colon cancer cells that are non-tumorigenic in nude mice; CRL-1459 (CCD-18CO) human colon endothelial cells that are non-malignant; CCL-220 (COLO 320DM) human colon cancer cells that are tumorigenic in nude mice; and HTB-171 (NCI H446) human lung carcinoma cells) were tested for their ability to bind to agarose beads derivatized with 51 different molecules. There were statistically significant differences in binding of the 4 cell types to all of the 51 types of beads, but 15 types of beads showed dramatic differences in binding to one or more of the 4 cell types. For example, only HTB-171 (NCI H446) bound to p-aminophenyl-beta-D-glucopyranoside-derivatized beads and only CCL-220 (COLO 320DM) bound to L-tyrosine-derivatized beads. The specificity of cell-bead binding was examined by performing assays in the presence or absence of exogenously added compounds in hapten-type of inhibition experiments. This assay, that utilizes large numbers of novel probes, may help in the development of new libraries of surface properties of specific cell types, with differing degrees of malignancy, that at this time could not be developed by using other available technologies.