MilliporeSigma
  • Recombinant β-Glucocerebrosidase specific immunoaffinity ligands selected from phage-displayed combinatorial scFv libraries.
Recombinant β-Glucocerebrosidase specific immunoaffinity ligands selected from phage-displayed combinatorial scFv libraries.
Protein expression and purification (2020-01-26)
R L Anisimov, O A Ershova, A V Ershov, M A Filatova, S A Katorkin, V M Simonov
ABSTRACT
Antibodies specific to β-Glucocerebrosidase were selected from phage displayed naïve scFv libraries. Biopannings were performed against recombinant human protein β-Glucocerebrosidase immobilized on polystyrene surface, specific phages were eluted with 50% ethylene glycol in citrate buffer (pH 6.0). Several specific binders were discovered and converted to full-size hIgG1 antibodies leading to highly stable binders with dissociation constants (Kd) in the range 10-40 nM. The antibodies were used further as ligands for affinity chromatography, where efficient and selective recovery of biologically active β-Glucocerebrosidase from cultured media of Chinese hamster ovary cells was demonstrated. β-Glucocerebrosidase was purified to nearly homogeneous state and had specific activity comparable to the commercially available preparations (40-44 U/mg protein). The obtained immunoaffinity sorbents have high capacity and can be easily regenerated.
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard