MilliporeSigma
  • CLIP-region mediated interaction of Invariant chain with MHC class I molecules.
CLIP-region mediated interaction of Invariant chain with MHC class I molecules.
FEBS letters (2006-05-09)
Simon J Powis
ABSTRACT
An association between the MHC class II chaperone molecule Invariant chain (Ii) and MHC class I molecules is known to occur, but the basis of the interaction is undetermined. Evidence is presented here that the CLIP region of Ii is involved in this interaction. A peptide encoding residues 91-99 of CLIP (MRMATPLLM) stabilised multiple MHC class I alleles, with the methionine residue at position 99 having a crucial role in binding to H2-K(b), whereas methionine at position 91 also appeared important in binding to RT1-A(a). Ii can also be detected in the class I MHC peptide loading complex. These data provide an explanation for the association of Ii and MHC class I molecules.