MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cranial and large vessel activity on positron emission tomography scan at diagnosis and 6 months in giant cell arteritis.
Cranial and large vessel activity on positron emission tomography scan at diagnosis and 6 months in giant cell arteritis.
International journal of rheumatic diseases (2020-02-27)
Anthony M Sammel, Edward Hsiao, Geoffrey Schembri, Elizabeth Bailey, Katherine Nguyen, Janice Brewer, Leslie Schrieber, Beatrice Janssen, Peter Youssef, Clare L Fraser, Rodger Laurent
ABSTRACT
Positron emission tomography/computed tomography (PET/CT) can detect cranial and large vessel inflammation in giant cell arteritis (GCA). We aimed to determine the change and significance of vascular activity at diagnosis and 6 months. Newly diagnosed GCA patients underwent time-of-flight fluorine-18-fluoro-2-deoxyglucose PET/CT from vertex to diaphragm within 72 hours of commencing corticosteroids and were followed for 12 months. A 6 months scan was performed in patients with inflammatory features on biopsy or CT aortitis. Vascular uptake was visually graded by 2 blinded readers across 18 artery segments from 0 (no increased uptake) to 3 (very marked uptake). Scores were summed to give a total vascular score (TVS). We enrolled 21 GCA patients and 15 underwent the serial scan. Twelve (57%) patients experienced a relapse and 5 of these had ischemic features of vision disturbance, jaw or limb claudication. The median TVS fell from 14 (interquartile range [IQR] 4-24) at baseline to 5 (IQR 0-10) at 6 months (P < .01) with reduction in both cranial and large artery scores. While the overall relapse rate was similar between patients with a high (≥10) and low baseline TVS, patients with high scores were numerically more likely to experience an ischemic relapse (33% vs 11%, P = .34). Five out of 15 patients had persistent uptake in at least 1 vessel on the serial PET/CT but none experienced a subsequent relapse. Vascular activity decreased in cranial and large arteries between diagnosis and 6 months. Persistent activity did not predict subsequent relapse.
Sigma-Aldrich
Golgicide A, ≥98% (HPLC)