MilliporeSigma
  • Comparison of the quality characteristics of chicken breast meat from conventional and animal welfare farms under refrigerated storage.
Comparison of the quality characteristics of chicken breast meat from conventional and animal welfare farms under refrigerated storage.
Poultry science (2020-03-01)
Hee-Jin Kim, Hye-Jin Kim, JinJoo Jeon, Ki-Chang Nam, Kwan-Seob Shim, Jong-Hyun Jung, Kyong Su Kim, Yangil Choi, Sang-Ho Kim, Aera Jang
ABSTRACT
In this study, we aimed to investigate the meat quality characteristics, bioactive compound content, and antioxidant activity during refrigerated storage of breast meat of Arbor Acres broilers (carcass weight: 1.1 kg, raised for 35 D) obtained from a conventional farm (BCF, n = 30) and an animal welfare farm (BAF, n = 30) in Korea. The BCF and BAF did not differ in their proximate composition, color, water-holding capacity, creatine, creatinine, and carnosine contents. However, the shear force value was significantly higher in BAF than in BCF (P < 0.05). The 2-thiobarbituric acid reactive substance (TBARS) levels in BCF on days 7 and 9 were significantly higher than those in BAF (P < 0.001). During storage, the total volatile basic nitrogen (VBN) content of BAF was significantly lower, except on day 1. The fatty acid composition of samples was not affected by the storage period, however, saturated fatty acid and unsaturated fatty acid contents did differ among the types of farm systems (P < 0.05). Although the creatine, creatinine, and carnosine contents in BAF and BCF did not differ significantly, the carnosine and creatinine contents decreased with the increase in storage period (P < 0.05). The anserine content of BAF was significantly higher than that of BCF throughout storage. Superoxide dismutase activity was not affected by the type of farm system but was affected by storage period. Overall, BAF showed lower pH, microorganism, TBARS, and VBN values, and higher anserine contents than BCF. These findings can serve as reference data for the evaluation of chicken meat quality of broilers raised in animal welfare farm and conventional farm.
Sigma-Aldrich
2-Methylheptane, 98%