MilliporeSigma
  • Cell-Selective Cytotoxicity of a Fluorescent Rhodium Metalloinsertor Conjugate Results from Irreversible DNA Damage at Base Pair Mismatches.
Cell-Selective Cytotoxicity of a Fluorescent Rhodium Metalloinsertor Conjugate Results from Irreversible DNA Damage at Base Pair Mismatches.
Biochemistry (2020-01-23)
Adela Nano, Julie M Bailis, Natalie F Mariano, Elizabeth D Pham, Stephanie D Threatt, Jacqueline K Barton
ABSTRACT
Up to 20% of solid tumors are characterized by DNA mismatch repair (MMR) deficiency and microsatellite instability that confer resistance to standard of care chemotherapy. MMR-deficient cancers have an increased mutation rate, and DNA mismatches accumulate as part of these cancers. We previously described a class of compounds, rhodium metalloinsertors, that bind DNA mismatches with high specificity and selectivity and have potential as targeted therapy. [Rh(chrysi)(phen)(PPO)]2+ (RhPPO) is the most potent, selective compound in this class and acts by targeting DNA mismatches, resulting in preferential cytotoxicity to MMR-deficient cancers. To explore further the cellular mechanism of action of RhPPO, we conjugated the metal complex to a fluorescent probe, cyanine 3 (Cy3). RhPPO-Cy3 binds DNA mismatches and retains the selectivity and potent cytotoxic activity of RhPPO for MMR-deficient cell lines. RhPPO-Cy3 forms discrete foci in the cell nucleus that overlap with sites of DNA damage, suggesting that the lesions occur at or near DNA mismatch sites. RhPPO-Cy3 foci persist over time, despite initial processing of the lesion and recruitment of repair proteins, consistent with the idea that the complex binding to a mismatch prevents repair. RhPPO-Cy3 binding does not lead to activation of p53 and the apoptotic pathway. Together, these findings support the idea that RhPPO-Cy3 binding leads to irreversible DNA damage at DNA mismatches that enables selective cytotoxicity to MMR-deficient cells.
Sigma-Aldrich
2,2′-Dipyridylamine, 98%
Sigma-Aldrich
Hexadecylphosphonic acid, 97%