MilliporeSigma
  • The mechanistic and functional profile of the therapeutic anti-IgE antibody ligelizumab differs from omalizumab.
The mechanistic and functional profile of the therapeutic anti-IgE antibody ligelizumab differs from omalizumab.
Nature communications (2020-01-09)
Pascal Gasser, Svetlana S Tarchevskaya, Pascal Guntern, Daniel Brigger, Rahel Ruppli, Noemi Zbären, Silke Kleinboelting, Christoph Heusser, Theodore S Jardetzky, Alexander Eggel
ABSTRACT
Targeting of immunoglobulin E (IgE) represents an interesting approach for the treatment of allergic disorders. A high-affinity monoclonal anti-IgE antibody, ligelizumab, has recently been developed to overcome some of the limitations associated with the clinical use of the therapeutic anti-IgE antibody, omalizumab. Here, we determine the molecular binding profile and functional modes-of-action of ligelizumab. We solve the crystal structure of ligelizumab bound to IgE, and report epitope differences between ligelizumab and omalizumab that contribute to their qualitatively distinct IgE-receptor inhibition profiles. While ligelizumab shows superior inhibition of IgE binding to FcεRI, basophil activation, IgE production by B cells and passive systemic anaphylaxis in an in vivo mouse model, ligelizumab is less potent in inhibiting IgE:CD23 interactions than omalizumab. Our data thus provide a structural and mechanistic foundation for understanding the efficient suppression of FcεRI-dependent allergic reactions by ligelizumab in vitro as well as in vivo.
Sigma-Aldrich
11-Amino-1-undecanethiol hydrochloride, 97%