MilliporeSigma
IDH1 mutations in gliomas: when an enzyme loses its grip.
Cancer cell (2010-02-05)
Christian Frezza, Daniel A Tennant, Eyal Gottlieb
ABSTRACT
The growing interest in cancer metabolism is best demonstrated by the rapid progress made in studying isocitrate dehydrogenase (IDH) mutations since their discovery just over a year ago. In a recent study published in Nature, Dang et al. identified 2-hydroxyglutarate as a product of tumor-associated IDH mutants with potential oncogenic activities.