MilliporeSigma
  • Rapid and profound rewiring of brain lipid signaling networks by acute diacylglycerol lipase inhibition.
Rapid and profound rewiring of brain lipid signaling networks by acute diacylglycerol lipase inhibition.
Proceedings of the National Academy of Sciences of the United States of America (2015-12-17)
Daisuke Ogasawara, Hui Deng, Andreu Viader, Marc P Baggelaar, Arjen Breman, Hans den Dulk, Adrianus M C H van den Nieuwendijk, Adriann M C H van den Nieuwendijk, Marjolein Soethoudt, Tom van der Wel, Juan Zhou, Herman S Overkleeft, Manuel Sanchez-Alavez, Simone Mori, Simone Mo, William Nguyen, Bruno Conti, Xiaojie Liu, Yao Chen, Qing-Song Liu, Benjamin F Cravatt, Mario van der Stelt
ABSTRACT
Diacylglycerol lipases (DAGLα and DAGLβ) convert diacylglycerol to the endocannabinoid 2-arachidonoylglycerol. Our understanding of DAGL function has been hindered by a lack of chemical probes that can perturb these enzymes in vivo. Here, we report a set of centrally active DAGL inhibitors and a structurally related control probe and their use, in combination with chemical proteomics and lipidomics, to determine the impact of acute DAGL blockade on brain lipid networks in mice. Within 2 h, DAGL inhibition produced a striking reorganization of bioactive lipids, including elevations in DAGs and reductions in endocannabinoids and eicosanoids. We also found that DAGLα is a short half-life protein, and the inactivation of DAGLs disrupts cannabinoid receptor-dependent synaptic plasticity and impairs neuroinflammatory responses, including lipopolysaccharide-induced anapyrexia. These findings illuminate the highly interconnected and dynamic nature of lipid signaling pathways in the brain and the central role that DAGL enzymes play in regulating this network.
Sigma-Aldrich
DO34, ≥98% (HPLC)