MilliporeSigma
  • Analysis of oak tannins by liquid chromatography-electrospray ionisation mass spectrometry.
Analysis of oak tannins by liquid chromatography-electrospray ionisation mass spectrometry.
Journal of chromatography. A (2000-09-22)
P Mämmelä, H Savolainen, L Lindroos, J Kangas, T Vartiainen
ABSTRACT
Extractable tannins were analysed by liquid chromatography-electrospray ionisation mass spectrometry in two oak species, North American white oak (Quercus alba) and European red oak (Quercus robur). They mainly included various glucose gallic and ellagic acid esters. The structures were partially determined, and they included grandinin/roburin E, castalagin/vescalagin, gallic acid, valoneic acid bilactone, monogalloyl glucose, digalloyl glucose, trigalloyl glucose, ellagic acid rhamnose, quercitrin and ellagic acid.