MilliporeSigma
  • Biodegradation of propyzamide by Comamonas testosteroni W1 and cloning of the propyzamide hydrolase gene camH.
Biodegradation of propyzamide by Comamonas testosteroni W1 and cloning of the propyzamide hydrolase gene camH.
Bioresource technology (2014-12-30)
Baiping Zhao, Xiude Hua, Fei Wang, Weiliang Dong, Zhoukun Li, Yu Yang, Zhongli Cui, Minghua Wang
ABSTRACT
Propyzamide is a widely used benzamide herbicide for controlling weeds in lettuce, soybeans, cotton and other crops. An efficient propyzamide-degrading strain W1 was firstly isolated from activated sludge and identified as Comamonas testosteroni. A metabolite of propyzamide by strain W1 was firstly identified. The novel gene camH encoding a hydrolase that catalyzed the amide bond cleavage of propyzamide was cloned from strain W1. The gene contained an open reading frame of 1452 bp, the deduced amino acid sequence showed low identity with other amidases. The recombinant enzyme CamH was expressed in Escherichia coli BL21 and purified. CamH displayed the highest activity at 30°C and pH 8.0 with propyzamide as the substrate. These results provide important knowledge on the fate of propyzamide in the biodegradation, and elucidate the biodegradation mechanism of propyzamide by the strain W1.