MilliporeSigma
  • A glycobiology review: carbohydrates, lectins and implications in cancer therapeutics.
A glycobiology review: carbohydrates, lectins and implications in cancer therapeutics.
Acta histochemica (2010-03-05)
Haike Ghazarian, Brian Idoni, Steven B Oppenheimer
ABSTRACT
This review is intended for general readers who would like a basic foundation in carbohydrate structure and function, lectin biology, and the implications of glycobiology in human health and disease, particularly in cancer therapeutics. These topics are among the hundreds included in the field of glycobiology and are treated here because they form the cornerstone of glycobiology or the focus of many advances in this rapidly expanding field.