MilliporeSigma
  • An efficient synthesis of polyguanylic acid by a thermophilic polynucleotide phosphorylase.
An efficient synthesis of polyguanylic acid by a thermophilic polynucleotide phosphorylase.
Biochimica et biophysica acta (1977-06-17)
Y Kikuchi, K Hirai, F Hishinuma, K Sakaguchi
PMID884104
ABSTRACT
Polyguanylic acid (poly(G)) was synthesized from GDP in a yield of 60-75% by Thermus thermophilus polynucleotide phosphorylase (polyribonucleotide: orthophosphate nucleotidyltransferase, EC 2.7.7.8) at 70 degrees C, pH 8.5 in the presence of Mg2+. The yield was dependent on the ratio of GDP to Mg2+, but was independent of the concentrations of enzyme or substrate. The maximal rate of GDP polymerization was obtained when the ratio of GDP to Mg2+ was 3:1. However, by prolonged incubation, the higher initial ratio of over 4:1 was preferred because of the rapid consumption of GDP in the reaction mixture. Poly(G) prepared by 1 h incubation was heterogeneous in size from 5 S to over 23 S, but by prolonged incubation of 19 h the size of product converged to 9 S as judged by sucrose density gradient centrifugation. Its chain length was determined by terminal nucleoside analysis to be 200 nucleotides long.
Sigma-Aldrich
Polyguanylic acid potassium salt, lyophilized powder