MilliporeSigma
  • Study on volatile components in salami by reverse carrier gas headspace gas chromatography-mass spectrometry.
Study on volatile components in salami by reverse carrier gas headspace gas chromatography-mass spectrometry.
Journal of chromatography. A (1999-02-19)
G Procida, L S Conte, S Fiorasi, G Comi, L G Favretto
ABSTRACT
Salami are a typical seasoned sausage of Italy; a number of types are produced, according to local traditional recipes. As industrial production has taken place, a number of problems rise in obtaining products similar to the traditional ones. The use of selected microbial starters is permitted by Italian law for some years and at present, microbiological research is engaged in selecting starters similar to the ones isolated from traditional products, with the aim of obtaining organoleptic characteristics close to the ones of traditional recipes. A study was carried out concerning the characterisation of volatile components of salami by headspace capillary gas chromatography-mass spectrometry. As during the sampling step, analytes could reach the analytical column, the carrier gas rate was back flushed in the latter, while a pre column was used as cold trap. Then GC-MS analysis follows. By these techniques, we were able to highlight typical profiles of different salami, as well as monitoring the ripening of a traditional and a starter added salami. Main peaks are of fermentative origin, while also peaks from spices were detected. Ethyl propionate was used as internal standard to be able to normalise the peaks amounts.