MilliporeSigma
BLU-285, DCC-2618 Show Activity against GIST.
Cancer discovery (2017-01-13)
ABSTRACT
Tyrosine kinase inhibitors may initially control gastrointestinal stromal tumors, but most patients eventually experience disease progression due to activation loop mutations, which are resistant to approved drugs. However, phase I trials suggest that the cancer is sensitive to two new agents: BLU-285 and DCC-2618. In addition, liquid biopsies, which were used to follow patients during the trials, may be valuable tools in tracking disease progression and assessing tumor heterogeneity.
Sigma-Aldrich
DCC-2618, ≥98% (HPLC)