MilliporeSigma
  • Fluorinated organocatalysts for the enantioselective epoxidation of enals: molecular preorganisation by the fluorine-iminium ion gauche effect.
Fluorinated organocatalysts for the enantioselective epoxidation of enals: molecular preorganisation by the fluorine-iminium ion gauche effect.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-26)
Eva-Maria Tanzer, Lucie E Zimmer, W Bernd Schweizer, Ryan Gilmour
ABSTRACT
The fluorine-iminium ion gauche effect is triggered upon union of a secondary β-fluoroamine and an α,β-unsaturated aldehyde, providing a useful strategy for controlling the molecular topology of intermediates that are central to organocatalytic processes. The β-fluoroamine (S)-2-(fluorodiphenylmethyl)pyrrolidine (1) is an effective catalyst for the enantioselective epoxidation of α,β-unsaturated aldehydes. A process of structural editing has revealed that the efficiency of this catalyst is due to the (fluorodiphenyl)methyl group when it is embedded in a β-fluoroiminium motif. Epoxidations of challenging cyclic α,β-disubstituted, β,β-disubstituted and α,β,β-trisubstituted enals catalysed by 1 proceed with excellent levels of enantiocontrol (up to 98% ee).