MilliporeSigma
  • Analysis of amino acids by high-performance liquid chromatography with circular dichroism detection
Analysis of amino acids by high-performance liquid chromatography with circular dichroism detection
Chemical & Pharmaceutical Bulletin (1995)
TAKATORI K, et al.