MilliporeSigma
  • Determination of the impurity profile of gamma-cyclodextrin by high-performance liquid chromatography
Determination of the impurity profile of gamma-cyclodextrin by high-performance liquid chromatography
Journal of Chromatography A (1992)
White G, et al.