MilliporeSigma
  • Tetrachloroethylene and trichloroethylene fatality: case report and simple headspace SPME-capillary gas chromatographic determination in tissues
Tetrachloroethylene and trichloroethylene fatality: case report and simple headspace SPME-capillary gas chromatographic determination in tissues
Journal of Analytical Toxicology (2000)
Dehon B, et al.