MilliporeSigma
  • VisG is eβential for biosynthesis of virginiamycin S, a streptogramin type B antibiotic, as a provider of the nonproteinogenic amino acid phenylglycine
VisG is eβential for biosynthesis of virginiamycin S, a streptogramin type B antibiotic, as a provider of the nonproteinogenic amino acid phenylglycine
Microbiology (2011)
Fitria Ningsih, Shigeru Kitani, et al.