MilliporeSigma
  • Short photoirradiation induces flavonoid synthesis and increases its production in postharvest vegetables.
Short photoirradiation induces flavonoid synthesis and increases its production in postharvest vegetables.
Journal of agricultural and food chemistry (2012-04-18)
Kazuki Kanazawa, Takashi Hashimoto, Satoko Yoshida, Park Sungwon, Shinya Fukuda
ABSTRACT
It is desirable to increase the flavonoid contents of postharvest vegetables since flavonoids play a beneficial role in human health promotion. In the present study, we show that postharvest vegetables increasingly produced flavonoids when irradiated with light near the absorption wavelength of flavonoids in the plant. Three-day exposure to UV-B for 5 min, 98 μmol m⁻² s⁻¹ per day, increased the contents of jaceidin in spinach, kaempherol glycoside in radish sprout, apigenin glycosides in parsley, and isovitexin in Indian spinach after 6 days of storage in a refrigerator, compared to the contents in plants without irradiation. Six days of storage of unripe green strawberry under green light for 5 min, 98 μmol m⁻² s⁻¹ per day, enabled them to mature and turn red, accompanied by 3.5-fold increased contents of pelargonidin. Elucidation of the mechanism in parsley found the stimulating expression of the flavonoid synthesis gene, PAL, C4H, 4CL, CHS, and FNS, 6 h after exposure to single irradiation with UV-B for 5 min, and the higher expression was maintained for 24 h. After 3 days irradiation during 6 days of storage, parsley did not show adverse changes in the contents of ascorbic acid, β-carotene, chlorophyll, and moisture.
Sigma-Aldrich
Isovitexin, ≥98.0% (HPLC)
Pricing and availability is not currently available.