MilliporeSigma
  • Fluorophore photostability and saturation in the hotspot of DNA origami nanoantennas.
Fluorophore photostability and saturation in the hotspot of DNA origami nanoantennas.
Methods and applications in fluorescence (2020-01-14)
Lennart Grabenhorst, Kateryna Trofymchuk, Florian Steiner, Viktorija Glembockyte, Philip Tinnefeld
ABSTRACT
Fluorescent dyes used for single-molecule spectroscopy can undergo millions of excitation-emission cycles before photobleaching. Due to the upconcentration of light in a plasmonic hotspot, the conditions for fluorescent dyes are even more demanding in DNA origami nanoantennas. Here, we briefly review the current state of fluorophore stabilization for single-molecule imaging and reveal additional factors relevant in the context of plasmonic fluorescence enhancement. We show that despite the improved photostability of single-molecule fluorophores by DNA origami nanoantennas, their performance in the intense electric fields in plasmonic hotspots is still limited by the underlying photophysical processes, such as formation of dim states and photoisomerization. These photophysical processes limit the photon count rates, increase heterogeneity and aggravate quantification of fluorescence enhancement factors. These factors also reduce the time resolution that can be achieved in biophysical single-molecule experiments. Finally, we show how the photophysics of a DNA hairpin assay with a fluorophore-quencher pair can be influenced by plasmonic DNA origami nanoantennas leading to implications for their use in fluorescence-based diagnostic assays. Especially, we show that such assays can produce false positive results by premature photobleaching of the dark quencher.
Sigma-Aldrich
N-Bromoacetamide, powder
Sigma-Aldrich
Potassium Phosphate Monobasic solution, reagent grade, 1.0 M in solution
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M