MilliporeSigma
  • Decomposing Mechanism of SF6 under Positive DC Partial Discharge in the Presence of Trace H2O.
Decomposing Mechanism of SF6 under Positive DC Partial Discharge in the Presence of Trace H2O.
ACS omega (2020-06-18)
Min Liu
ABSTRACT
The influence of H2O on SF6 decomposition characteristics under positive DC partial discharge (PD) is significant. To evaluate PD fault severity in DC SF6-insulated equipment using the production characteristics of SF6 decomposition components, the corresponding relationship and mathematical expression between the production of SF6 decomposition components and the H2O content should be identified and achieved. Thus, SF6 decomposition experiments under positive DC PD are performed to reflect the influence of H2O on SF6 decomposition components. Results show that the total discharge quantity and the discharge repetition rate averaged for 1 s decrease slightly when the H2O content increases from 0 to 970 ppmv and then increase when the H2O content increases from 970 to 5120 ppmv. The effective production rates of SO2F2, SOF2, and SO2 increase with the H2O content, whereas that of SOF4 decreases. Finally, the corresponding relationship and mathematical expression between the characteristic ratio (c(SO2F2) + c(SOF4))/(c(SOF2) + c(SO2)) of components and the H2O content have been achieved, which can afford references for PD fault diagnosis in DC SF6 gas-insulated equipment.