MilliporeSigma
  • Development of new semiconducting polymers for high performance solar cells.
Development of new semiconducting polymers for high performance solar cells.
Journal of the American Chemical Society (2008-12-20)
Yongye Liang, Yue Wu, Danqin Feng, Szu-Ting Tsai, Hae-Jung Son, Gang Li, Luping Yu
ABSTRACT
A new low band gap semiconducting polymer, PTB1, was synthesized and found promising for solar energy harvesting. Simple polymer solar cells based on PTB1 and methanofullerene [6,6]-phenyl-C(71)-butyric acid methyl esters (PC(71)BM) exhibit a solar conversion efficiency of 5.6%. An external quantum efficiency of 67% and fill-factor of 65% are achieved, both of which are among the highest values reported for a solar cell system based on a low band gap polymer.