MilliporeSigma
  • Exploring SnxTi1-xO2 Solid Solutions Grown onto Graphene Oxide (GO) as Selective Toluene Gas Sensors.
Exploring SnxTi1-xO2 Solid Solutions Grown onto Graphene Oxide (GO) as Selective Toluene Gas Sensors.
Nanomaterials (Basel, Switzerland) (2020-04-25)
Eleonora Pargoletti, Simone Verga, Gian Luca Chiarello, Mariangela Longhi, Giuseppina Cerrato, Alessia Giordana, Giuseppe Cappelletti
ABSTRACT
The major drawback of oxide-based sensors is the lack of selectivity. In this context, SnxTi1-xO2/graphene oxide (GO)-based materials were synthesized via a simple hydrothermal route, varying the titanium content in the tin dioxide matrix. Then, toluene and acetone gas sensing performances of the as-prepared sensors were systematically investigated. Specifically, by using 32:1 SnO2/GO and 32:1 TiO2/GO, a greater selectivity towards acetone analyte, also at room temperature, was obtained even at ppb level. However, solid solutions possessing a higher content of tin relative to titanium (as 32:1 Sn0.55Ti0.45O2/GO) exhibited higher selectivity towards bigger and non-polar molecules (such as toluene) at 350 °C, rather than acetone. A deep experimental investigation of structural (XRPD and Raman), morphological (SEM, TEM, BET surface area and pores volume) and surface (XPS analyses) properties allowed us to give a feasible explanation of the different selectivity. Moreover, by exploiting the UV light, the lowest operating temperature to obtain a significant and reliable signal was 250 °C, keeping the greater selectivity to the toluene analyte. Hence, the feasibility of tuning the chemical selectivity by engineering the relative amount of SnO2 and TiO2 is a promising feature that may guide the future development of miniaturized chemoresistors.
Sigma-Aldrich
Tin(IV) chloride pentahydrate, 98%