MilliporeSigma
  • Non-basic high-performance molecules for solution-processed organic solar cells.
Non-basic high-performance molecules for solution-processed organic solar cells.
Advanced materials (Deerfield Beach, Fla.) (2012-06-08)
Thomas S van der Poll, John A Love, Thuc-Quyen Nguyen, Guillermo C Bazan
ABSTRACT
A new small molecule, p-DTS(FBTTh(2))(2), is designed for incorporation into solution-fabricated high-efficiency organic solar cells. Of primary importance is the incorporation of electron poor heterocycles that are not prone to protonation and thereby enable the incorporation of commonly used interlayers between the organic semiconductor and the charge collecting electrodes. These features have led to the creation of p-DTS(FBTTh(2))(2)/PC(71)BM solar cells with power conversion efficiencies of up to 7%.