MilliporeSigma
  • Circular RNA circHIPK3 modulates the proliferation of airway smooth muscle cells by miR-326/STIM1 axis.
Circular RNA circHIPK3 modulates the proliferation of airway smooth muscle cells by miR-326/STIM1 axis.
Life sciences (2020-05-26)
Junling Lin, Xiaokai Feng, Jun Zhang
ABSTRACT
Emerging findings demonstrate the critical roles of noncoding RNA (ncRNA) in asthma development. Nevertheless, the biological roles of circular RNA (circRNA) in airway remodeling are still elusive. Here, the present research focuses on the regulation of circRNA circHIPK3 in airway smooth muscle cells (ASMCs) proliferation and migration. The sequence of circRNA was detected using Sanger sequencing. Cellular phenotypes were detected using CCK-8 assay, transwell and flow cytometer assay. The potential binding of miRNA and downstream and upstream targets was detected using dual-luciferase reporter assay. Results showed that circHIPK3 was significantly upregulated in platelet-derived growth factor (PDGF) induced ASMCs. Functional analysis using CCK-8, transwell migration assays and flow cytometry analysis showed that circHIPK3 knockdown repressed proliferation, migration and up-regulated the apoptosis in ASMCs. Mechanistic assays showed that circHIPK3 sponged miR-326 in the cytoplasm, thereby targeting stromal interaction molecule 1 (STIM1) to regulate ASMCs' proliferation, migration and apoptosis. Collectively, the data elucidates that circHIPK3 functions as a regulator in the airway remodeling during the asthma development through miR-326/STIM1 axis, providing a novel insight for the therapeutic target.
Sigma-Aldrich
MISSION® esiRNA, targeting human HIPK3