MilliporeSigma
  • Neutralizing antibody against osteopontin attenuates non-alcoholic steatohepatitis in mice.
Neutralizing antibody against osteopontin attenuates non-alcoholic steatohepatitis in mice.
Journal of cell communication and signaling (2020-02-18)
Machiko Honda, Chiemi Kimura, Toshimitsu Uede, Shigeyuki Kon
ABSTRACT
Previously, we reported that an extracellular matrix protein, osteopontin (OPN), is involved in various autoimmune diseases using a neutralizing polyclonal antibody against OPN generated in rabbits. However, the antibody cannot be used for long-term mouse models of chronic inflammatory disease because of the induction of antibodies against anti-OPN rabbit IgG. In this study, we generated a new antibody, anti-mouse OPN mouse IgG (35B6). 35B6 inhibited the cell adhesion of mouse and human OPN to Chinese Hamster Ovary (CHO) cells or CHO cells expressing α4 or α9 integrin. It was reported that OPN is highly expressed and has an important role in a chronic liver disease, non-alcoholic steatohepatitis (NASH). 35B6 injection twice a week for 8 weeks attenuated liver inflammation and fibrosis in a NASH mouse model, suggesting 35B6 is beneficial for the treatment of NASH. 35B6 was preferable to the rabbit anti-OPN antibody for investigating the in vivo function of OPN in mouse models of long-term disease.
Sigma-Aldrich
Rat OPN ELISA Kit, for cell culture supernatants, plasma, and serum samples