MilliporeSigma
  • A Glycosaminoglycan-Rich Fraction from Sea Cucumber Isostichopus badionotus Has Potent Anti-Inflammatory Properties In Vitro and In Vivo.
A Glycosaminoglycan-Rich Fraction from Sea Cucumber Isostichopus badionotus Has Potent Anti-Inflammatory Properties In Vitro and In Vivo.
Nutrients (2020-06-11)
Leticia Olivera-Castillo, George Grant, Nuvia Kantún-Moreno, Hirian A Barrera-Pérez, Jorge Montero, Miguel A Olvera-Novoa, Leydi M Carrillo-Cocom, Juan J Acevedo, Cesar Puerto-Castillo, Victor May Solís, Juan A Pérez-Vega, Judit Gil-Zamorano, Enrique Hernández-Garibay, María A Fernández-Herrera, Mayra Pérez-Tapia, Oscar Medina-Contreras, Jairo R Villanueva-Toledo, Rossanna Rodriguez-Canul, Alberto Dávalos
ABSTRACT
Sea cucumber body wall contains several naturally occurring bioactive components that possess health-promoting properties. Isostichopus badionotus from Yucatan, Mexico is heavily fished, but little is known about its bioactive constituents. We previously established that I. badionotus meal had potent anti-inflammatory properties in vivo. We have now screened some of its constituents for anti-inflammatory activity in vitro. Glycosaminoglycan and soluble protein preparations reduced 12-O-tetradecanoylphorbol-13-acetate (TPA)-induced inflammatory responses in HaCaT cells while an ethanol extract had a limited effect. The primary glycosaminoglycan (fucosylated chondroitin sulfate; FCS) was purified and tested for anti-inflammatory activity in vivo. FCS modulated the expression of critical genes, including NF-ĸB, TNFα, iNOS, and COX-2, and attenuated inflammation and tissue damage caused by TPA in a mouse ear inflammation model. It also mitigated colonic colitis caused in mice by dextran sodium sulfate. FCS from I. badionotus of the Yucatan Peninsula thus had strong anti-inflammatory properties in vivo.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder