MilliporeSigma
  • Composition of amino acids and related nitrogenous nutrients in feedstuffs for animal diets.
Composition of amino acids and related nitrogenous nutrients in feedstuffs for animal diets.
Amino acids (2020-03-13)
Peng Li, Guoyao Wu
ABSTRACT
We analyzed the composition of amino acids (AAs) in oligopeptides, proteins, and the free pool, as well as creatine, agmatine, polyamines, carnosine, anserine, and glutathione, in animal- and plant-derived feedstuffs. Ingredients of animal origins were black soldier fly larvae meal (BSFM), chicken by-product meal, chicken visceral digest, feather meal, Menhaden fishmeal, Peruvian anchovy fishmeal, Southeast Asian fishmeal, spray-dried peptone from enzymes-treated porcine mucosal tissues, poultry by-product meal (pet-food grade), spray-dried poultry plasma, and spray-dried egg product. Ingredients of plant origins were algae spirulina meal, soybean meal, and soy protein concentrate. All animal-derived feedstuffs contained large amounts of all proteinogenic AAs (particularly glycine, proline, glutamate, leucine, lysine, and arginine) and key nonproteinogenic AAs (taurine and 4-hydroxyproline), as well as significant amounts of agmatine, polyamines, creatine, creatinine, creatine phosphate, and glutathione. These nitrogenous substances are essential to either DNA and protein syntheses in cells or energy metabolism in tissues (particularly the brain and skeletal muscle). Of note, chicken by-product meal, poultry by-product meal, and spray-dried poultry plasma contained large amounts of carnosine and anserine (potent antioxidants). Compared with most of the animal-derived feedstuffs, plant-derived feedstuffs contained much lower contents of glycine and proline, little 4-hydroxyproline, and no creatine, creatinine, creatine phosphate, carnosine or anserine. These results indicate the unique importance of animal-source feedstuffs in improving the feed efficiency, growth and health of animals (including fish and companion animals). Because soy protein concentrate is consumed by infants, children and adults, as are BSFM and algae for children and adults, our findings also have important implications for human nutrition.
Sigma-Aldrich
Prolidase from porcine kidney, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder