MilliporeSigma
  • Fabrication of High-Performance Bamboo-Plastic Composites Reinforced by Natural Halloysite Nanotubes.
Fabrication of High-Performance Bamboo-Plastic Composites Reinforced by Natural Halloysite Nanotubes.
Molecules (Basel, Switzerland) (2020-05-15)
Xiaobei Jin, Jingpeng Li, Rong Zhang, Zehui Jiang, Daochun Qin
ABSTRACT
Bamboo-plastic composites (BPCs) as new biomass-plastic composites have recently attracted much attention. However, weak mechanical performance and high moisture absorption as well as low thermal stability greatly limit their industrial applications. In this context, different amounts of halloysite nanotubes (HNTs) were used as a natural reinforcing filler for BPCs. It was found that the thermal stability of BPCs increased with increasing HNT contents. The mechanical strength of BPCs was improved with the increase in HNT loading up to 4 wt% and then worsened, while the impact strengths were slightly reduced. Low HNT content (below 4 wt%) also improved the dynamic thermomechanical properties and reduced the water absorption of the BPCs. Morphological studies confirmed the improved interfacial compatibility of the BPC matrix with 4 wt% HNT loading, and high-concentration HNT loading (above 6 wt%) resulted in easy agglomeration. The results highlight that HNTs could be a feasible candidate as nanoreinforcements for the development of high-performance BPCs.
Sigma-Aldrich
N-Hydroxynaphthalimide triflate, electronic grade, ≥99%