MilliporeSigma
  • Xenogeneic Neu5Gc and self-glycan Neu5Ac epitopes are potential immune targets in MS.
Xenogeneic Neu5Gc and self-glycan Neu5Ac epitopes are potential immune targets in MS.
Neurology(R) neuroimmunology & neuroinflammation (2020-02-06)
Kayluz F Boligan, Johanna Oechtering, Christian W Keller, Benjamin Peschke, Robert Rieben, Nicolai Bovin, Ludwig Kappos, Richard D Cummings, Jens Kuhle, Stephan von Gunten, Jan D Lünemann
ABSTRACT
To explore the repertoire of glycan-specific immunoglobulin G (IgG) antibodies in treatment-naive patients with relapsing-remitting multiple sclerosis (RRMS). A systems-level approach combined with glycan array technologies was used to determine specificities and binding reactivities of glycan-specific IgGs in treatment-naive patients with RRMS compared with patients with noninflammatory and other inflammatory neurologic diseases. We identified a unique signature of glycan-binding IgG in MS with high reactivities to the dietary xenoglycan N-glycolylneuraminic acid (Neu5Gc) and the self-glycan N-acetylneuraminic acid (Neu5Ac). Increased reactivities of serum IgG toward Neu5Gc and Neu5Ac were additionally observed in an independent, treatment-naive cohort of patients with RRMS. Patients with MS show increased IgG reactivities to structurally related xenogeneic and human neuraminic acids. The discovery of these glycan-specific epitopes as immune targets and potential biomarkers in MS merits further investigation.