MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Oxidative damage mediates the association between polycyclic aromatic hydrocarbon exposure and lung function.
Oxidative damage mediates the association between polycyclic aromatic hydrocarbon exposure and lung function.
Environmental health : a global access science source (2020-07-04)
Limin Cao, Yun Zhou, Aijun Tan, Tingming Shi, Chunmei Zhu, Lili Xiao, Zhuang Zhang, Shijie Yang, Ge Mu, Xing Wang, Dongming Wang, Jixuan Ma, Weihong Chen
ABSTRACT
Exposure to polycyclic aromatic hydrocarbons (PAHs) is related to decreased lung function. However, whether oxidative damage is involved in this relationship remains unclear. This study was aimed to explore the potential mediating role of oxidative DNA or lipid damage in the association between PAH exposure and lung function. The urinary levels of monohydroxy polycyclic aromatic hydrocarbon metabolites (OH-PAHs) and lung function parameters were measured among 3367 participants from the baseline of the Wuhan-Zhuhai cohort. Urinary 8-hydroxy-2'-deoxyguanosine (8-OHdG) and 8-isoprostane (8-iso-PGF2α) were determined to evaluate the individuals' oxidative DNA and lipid damage degrees, respectively. Linear mixed models were used to investigate the associations of urinary OH-PAHs, 8-OHdG and 8-iso-PGF2α with lung function parameters. Mediation analysis was further conducted to assess the potential role of oxidative damage in the association between urinary OH-PAHs and lung function. Each one-percentage increase in the sum of urinary OH-PAHs, high-molecular-weight or low-molecular-weight OH-PAHs (ƩOH-PAHs, ƩHMW OH-PAH or ƩLMW OH-PAHs, respectively) was associated with a 0.2152-, 0.2076- or 0.1985- ml decrease in FEV1, and a 0.1891-, 0.2195- or 0.1634- ml decrease in FVC, respectively. Additionally, significantly positive dose-response relationships of ƩOH-PAHs, ƩHMW OH-PAH and ƩLMW OH-PAHs with urinary 8-OHdG or 8-iso-PGF2α, as well as an inverse dose-response relationship between urinary 8-OHdG and FVC, were observed (all P for trend < 0.05). Mediation analysis indicated that urinary 8-OHdG mediated 14.22% of the association between ƩHMW OH-PAH and FVC. Higher levels of oxidative DNA damage might be involved in the decreased levels of FVC caused by high-molecular-weight PAH exposure.
Sigma-Aldrich
9-Phenanthrol, technical grade