MilliporeSigma
  • Upregulation of PUM1 Expression in Preeclampsia Impairs Trophoblast Invasion by Negatively Regulating the Expression of the lncRNA HOTAIR.
Upregulation of PUM1 Expression in Preeclampsia Impairs Trophoblast Invasion by Negatively Regulating the Expression of the lncRNA HOTAIR.
Molecular therapy : the journal of the American Society of Gene Therapy (2019-12-22)
Yan Zhang, Xiao-Ying He, Shi Qin, Hui-Qin Mo, Xiao Li, Fan Wu, Jing Zhang, Xing Li, Lin Mao, Ya-Qing Peng, Yu-Na Guo, Yi Lin, Fu-Ju Tian
ABSTRACT
Pumilio (PUM) proteins are members of a highly conserved RNA-binding protein family that posttranscriptionally regulate gene expression in many organisms. However, their roles in the placenta are unclear. In the present study, we report the requirement for the PUM homolog 1 (PUM1) gene in preeclampsia (PE). Immunofluorescence and immunohistochemical data showed that PUM1 was highly expressed in human placental villi from women with PE compared to healthy controls (HCs). Further, PUM1 overexpression repressed, and knockdown enhanced, the invasion and proliferation of trophoblasts. Interestingly, PUM1 knockdown promoted trophoblast invasion in a villous explant culture model, while PUM1 overexpression repressed these effects. Furthermore, lncRNA transcriptome sequencing coupled with RNA immunoprecipitation (RIP) revealed that PUM1 inhibits trophoblast invasion in PE by downregulating the expression of lncRNA HOTAIR. Moreover, PUM1 regulates HOTAIR expression via a posttranscriptional mechanism. Using RNA-protein pull-down and mRNA stability assays, we identified PUM1 as a specific binding partner that decreased the half-life of HOTAIR and lowered the steady-state level of HOTAIR expression, suggesting a novel posttranscriptional regulatory mechanism. Collectively, these findings identified a novel RNA regulatory mechanism, revealing a new pathway governing the regulation of PUM1/HOTAIR in trophoblast invasion in the pathogenesis of PE.